Assay Details
| Gene Symbol: MIR5092 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049815.1 | 1 | 49 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 12 member 8 | SLC12A8 | CCC9 | Sense | Chr. 3 - 125082636 - 125212765 on NCBI Build GRCh38 | |
| Downstream | zinc finger protein 148 | ZNF148 |
BERF-1, BFCOL1, HT-BETA, ZBP-89, ZFP148, pHZ-52 |
74.1kb | Sense | Chr. 3 - 125225669 - 125375354 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0300012244.hg.1
Set Membership: |
|

MIR5092
Genomic Map