Assay Details
| Gene Symbol: MIR5190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049822.1 | 1 | 41 | 53 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LDLRAD4 antisense RNA 1 | LDLRAD4-AS1 | -32.4kb | Anti-sense | Chr. 18 - 13419499 - 13427534 on NCBI Build GRCh38 | |
| Downstream | microRNA 4526 | MIR4526 | mir-4526 | 151.1kb | Sense | Chr. 18 - 13611114 - 13611200 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1800006768.hg.1
Set Membership: |
|

MIR5190
Genomic Map