Assay Details
| Gene Symbol: MIR5587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049852.1 | 1 | 28 | 51 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 235 | LINC00235 |
C16orf10, LA16c-356B8.1, NCRNA00235 |
-7.9kb | Anti-sense | Chr. 16 - 526847 - 527407 on NCBI Build GRCh38 |
| Downstream | microRNA 3176 | MIR3176 | mir-3176 | 7.9kb | Sense | Chr. 16 - 543277 - 543366 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1600006469.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | ENST00000581803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006469.hg.1 | NR_049852.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600144505.hg.1 | ENST00000581803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600144505.hg.1 | NR_049852.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600144505.hg.1 | ENST00000581803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600144505.hg.1 | NR_049852.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600144505.hg.1 | TC1600006469.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000219611.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000562370.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000565010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000566977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000567216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568402.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000568988.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | ENST00000581803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NM_005632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006469.hg.1 | NR_049852.1 | Refseq |

MIR5587
Genomic Map