Assay Details
| Gene Symbol: MIR5683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049863.1 | 1 | 38 | 75 | ||
| Gene Symbol: MIR7853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_107007.1 | 1 | 75 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuritin 1 | NRN1 |
NRN, dJ380B8.2 |
-161.7kb | Anti-sense | Chr. 6 - 5997999 - 6007605 on NCBI Build GRCh38 |
| Downstream | LY86 antisense RNA 1 | LY86-AS1 |
LY86-AS, LY86AS |
177.1kb | Anti-sense | Chr. 6 - 6346465 - 6622826 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0600006824.hg.1, TC0600010710.hg.1
Set Membership: |
|

MIR5683
Genomic Map