Assay Details
| Gene Symbol: MIR5684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049864.1 | 1 | 28 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hook microtubule-tethering protein 2 | HOOK2 | HK2 | -8.6kb | Anti-sense | Chr. 19 - 12763002 - 12778484 on NCBI Build GRCh38 |
| Downstream |
JunB proto-oncogene, AP-1 transcription factor subunit |
JUNB | AP-1 | 4.3kb | Sense | Chr. 19 - 12791496 - 12793311 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1900007095.hg.1
Set Membership: |
|

MIR5684
Genomic Map