Assay Details
| Gene Symbol: MIR4666B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049877.1 | 1 | 40 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6134 | MIR6134 | hsa-mir-6134 | -1.1mb | Anti-sense | Chr. X - 28495555 - 28495663 on NCBI Build GRCh38 |
| Downstream | MAGE family member B2 | MAGEB2 |
CT3.2, DAM6, MAGE-XP-2 |
641.2kb | Sense | Chr. X - 30215558 - 30220089 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0X00006871.hg.1
Set Membership: |
|

MIR4666B
Genomic Map