Assay Details
| Gene Symbol: MIR5693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049878.1 | 1 | 36 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serpin family E member 3 | SERPINE3 | Anti-sense | Chr. 13 - 51334976 - 51362391 on NCBI Build GRCh38 | ||
| Downstream | integrator complex subunit 6 | INTS6 |
DBI-1, DDX26, DDX26A, DICE1, HDB, INT6, Notchl2 |
5.2kb | Sense | Chr. 13 - 51353171 - 51453236 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1300008982.hg.1
Set Membership: |
|

MIR5693
Genomic Map