Assay Details
Gene Symbol: LINC00534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_051989.1 | 1 - 2 | 261 | 64 | ||
GenBank mRNA | BG492574.1 | - | 1 - 2 | 212 | 64 | |
BG494657.1 | - | 2 - 3 | 124 | 64 | ||
BG496346.1 | - | 1 - 2 | 220 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156234.hg.1 | TC0800008186.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156232.hg.1 | TC0800008186.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156233.hg.1 | TC0800008186.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | NR_051989.1 | Refseq |
