Assay Details
| Gene Symbol: UBE2W | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073120.1 | 5 - 6 | 506 | 110 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | staufen double-stranded RNA binding protein 2 | STAU2 |
39K2, 39K3 |
-38.5kb | Sense | Chr. 8 - 73420369 - 73747708 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1617 | LINC01617 |
C8orf23, NCRNA00249, XLOC_006844 |
82.3kb | Anti-sense | Chr. 8 - 73879010 - 73904730 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

UBE2W
Genomic Map