Assay Details
| Gene Symbol: NEDD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142393.1 | NP_001135865.1 | 4 - 5 | 921 | 94 | |
| NM_001271033.1 | NP_001257962.1 | 2 - 3 | 279 | 94 | ||
| NM_006403.3 | NP_006394.1 | 3 - 4 | 726 | 94 | ||
| NR_073131.1 | 6 - 7 | 1166 | 94 | |||
| GenBank mRNA | AK292682.1 | - | 4 - 5 | 921 | 94 | |
| BC040207.1 | - | 3 - 4 | 712 | 4820745 | 94 | |
| BP262776.1 | - | 2 - 3 | 260 | 94 | ||
| BX648041.1 | - | 6 - 7 | 1133 | 94 | ||
More Information
Set Membership: |
|

NEDD9
Genomic Map