Assay Details
| Gene Symbol: PGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073143.2 | 4 - 5 | 2048 | 88 | ||
| XM_006718858.3 | XP_006718921.1 | 5 - 6 | 3954 | 88 | ||
| GenBank mRNA | AB085683.1 | - | 4 - 5 | 2055 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 133 | TMEM133 | AD031 | -48.0kb | Anti-sense | Chr. 11 - 100992080 - 100993935 on NCBI Build GRCh38 |
| Downstream | progesterone receptor antisense RNA | LOC101054525 | 77.5kb | Anti-sense | Chr. 11 - 101129077 - 101159270 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

PGR
Genomic Map