Assay Details
| Gene Symbol: MIR1302-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031633.1 | 1 | 49 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927865 | LOC101927865 | -15.0kb | Sense | Chr. 2 - 207186690 - 207254316 on NCBI Build GRCh38 | |
| Downstream | cAMP responsive element binding protein 1 | CREB1 |
CREB, CREB-1 |
260.5kb | Anti-sense | Chr. 2 - 207529892 - 207605989 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0200015544.hg.1
Set Membership: |
|

MIR1302-4
Genomic Map