Assay Details
| Gene Symbol: SNORA70D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033337.1 | 1 | 78 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MARVEL domain containing 3 | MARVELD3 |
MARVD3, MRVLDC3 |
-56.6kb | Anti-sense | Chr. 16 - 71626137 - 71641965 on NCBI Build GRCh38 |
| Downstream | adaptor related protein complex 1 gamma 1 subunit | AP1G1 |
ADTG, CLAPG1 |
30.3kb | Sense | Chr. 16 - 71729000 - 71809073 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001251.hg.1
Set Membership: |
|

SNORA70D
Genomic Map