Assay Details
| Gene Symbol: MIR5194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049826.1 | 1 | 53 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gasdermin C | GSDMC | MLZE | -221.4kb | Sense | Chr. 8 - 129705903 - 129786888 on NCBI Build GRCh38 |
| Downstream |
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1 |
ASAP1 |
AMAP1, CENTB4, DDEF1, PAG2, PAP, ZG14P |
43.7kb | Sense | Chr. 8 - 130052104 - 130443848 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0800011823.hg.1
Set Membership: |
|

MIR5194
Genomic Map