Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029440.hg.1 |
TC01001888.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029440.hg.1 |
ENST00000284548.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029440.hg.1 |
ENST00000366707.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029440.hg.1 |
ENST00000366709.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029440.hg.1 |
ENST00000422127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029440.hg.1 |
NM_001098623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029440.hg.1 |
NM_052843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
ENST00000284548.15 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
ENST00000366707.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
ENST00000422127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
ENST00000570156.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
NM_001098623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
NM_001271223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
NM_052843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
ENST00000284548.15 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
ENST00000366707.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
ENST00000422127.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
ENST00000570156.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
NM_001098623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
NM_001271223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
NM_052843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187603.hg.1 |
TC0100011861.hg.1 |
Clariom D TC |