Assay Details
| Gene Symbol: ST3GAL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271142.1 | NP_001258071.1 | 9 - 10 | 1135 | 109 | |
| NM_001271145.1 | NP_001258074.1 | 8 - 9 | 1076 | 109 | ||
| NM_001271146.1 | NP_001258075.1 | 8 - 9 | 1065 | 109 | ||
| NM_001271147.1 | NP_001258076.1 | 8 - 9 | 1039 | 109 | ||
| NM_001271148.1 | NP_001258077.1 | 5 - 6 | 823 | 109 | ||
| NM_001323352.1 | NP_001310281.1 | 9 - 10 | 1237 | 109 | ||
| NM_001323353.1 | NP_001310282.1 | 7 - 8 | 1001 | 109 | ||
| NM_001323358.1 | NP_001310287.1 | 10 - 11 | 1246 | 109 | ||
| NM_001323359.1 | NP_001310288.1 | 9 - 10 | 1146 | 109 | ||
| NM_001323360.1 | NP_001310289.1 | 10 - 11 | 1307 | 109 | ||
| NM_001323362.1 | NP_001310291.1 | 8 - 9 | 1173 | 109 | ||
| NM_001323363.1 | NP_001310292.1 | 7 - 8 | 1012 | 109 | ||
| NM_001323364.1 | NP_001310293.1 | 8 - 9 | 1184 | 109 | ||
| NM_001323365.1 | NP_001310294.1 | 9 - 10 | 1076 | 109 | ||
| NM_001323366.1 | NP_001310295.1 | 10 - 11 | 1146 | 109 | ||
| NM_001323367.1 | NP_001310296.1 | 8 - 9 | 893 | 109 | ||
| NM_001323368.1 | NP_001310297.1 | 8 - 9 | 901 | 109 | ||
| NM_006100.3 | NP_006091.1 | 9 - 10 | 1248 | 109 | ||
| GenBank mRNA | AB022918.1 | - | 8 - 9 | 1015 | 109 | |
| AF119391.1 | - | 8 - 9 | 1059 | 109 | ||
| AK001922.1 | - | 8 - 9 | 826 | 109 | ||
| AK315111.1 | - | 9 - 10 | 1239 | 109 | ||
| AL539506.3 | - | 5 - 6 | 680 | 109 | ||
| BC001266.2 | - | 9 - 10 | 1063 | 109 | ||
| BC023312.1 | - | 9 - 10 | 1224 | 4996181 | 109 | |
More Information
Set Membership: |
|

ST3GAL6
Genomic Map