Assay Details
| Gene Symbol: GZMH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033423.4 | NP_219491.1 | 4 - 5 | 710 | 118 | |
| XM_011536683.2 | XP_011534985.1 | 4 - 5 | 766 | 118 | ||
| GenBank mRNA | AW204493.1 | - | 1 - 2 | 118 | ||
| BC027974.1 | - | 4 - 5 | 648 | 5229314 | 118 | |
| BQ054303.1 | - | 4 - 5 | 710 | 118 | ||
| M36118.1 | - | 4 - 5 | 655 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000981.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14011256.hg.1 | ENST00000216338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14011256.hg.1 | ENST00000382547.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14011256.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14011256.hg.1 | ENST00000557220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14011256.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14011256.hg.1 | TC14000981.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164568.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164568.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164568.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164568.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164568.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164568.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164568.hg.1 | TC1400008763.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164565.hg.1 | TC1400008763.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008763.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164567.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164567.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164567.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164567.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164567.hg.1 | TC1400008763.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | ENST00000216338.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | ENST00000382548.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | ENST00000557220.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | NM_001270780.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | NM_001270781.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | NM_033423.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164566.hg.1 | TC1400008763.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 704559 | - | - |

GZMH
Genomic Map