Assay Details
| Gene Symbol: THEGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256475.1 | NP_001243404.1 | 8 - 9 | 1351 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000510716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007597.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152001.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152001.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152001.hg.1 | ENST00000512175.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152001.hg.1 | NM_001256475.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152001.hg.1 | TC0400007597.hg.1 | Clariom D TC |

THEGL
Genomic Map