Assay Details
| Gene Symbol: NTAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270766.1 | NP_001257695.1 | 6 - 7 | 545 | 80 | |
| NM_001270767.1 | NP_001257696.1 | 5 - 6 | 479 | 80 | ||
| NM_173474.3 | NP_775745.1 | 7 - 8 | 648 | 80 | ||
| XM_011522355.2 | XP_011520657.1 | 4 - 5 | 483 | 80 | ||
| GenBank mRNA | AF092440.1 | - | 7 - 8 | 555 | 80 | |
| AK092915.1 | - | 3 - 4 | 1831 | 80 | ||
| BC017336.2 | - | 7 - 8 | 545 | 4872717 | 80 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NTAN1
Genomic Map