| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007905.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900158929.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158929.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158929.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900158929.hg.1 |
TC1900007905.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900158929.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158929.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158929.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007905.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900158927.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158927.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158927.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900158927.hg.1 |
TC1900007905.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900158927.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158927.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158927.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900158931.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158931.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158931.hg.1 |
NM_001302757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900158931.hg.1 |
TC1900007905.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900158931.hg.1 |
ENST00000585332.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158931.hg.1 |
ENST00000633214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900158931.hg.1 |
NM_001302757.1 |
Refseq |