Assay Details
| Gene Symbol: MLL-MLL fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY187943.1 | - | 150 | 133 | ||
| AY187945.1 | - | 279 | 265 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLL-MLL fusion| Gene Symbol: MLL-MLL fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY187943.1 | - | 150 | 133 | ||
| AY187945.1 | - | 279 | 265 | |||
Set Membership: |
|