Assay Details
| Gene Symbol: IL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012631.1 | NP_001012649.1 | 1 - 2 | 182 | 77 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001012631.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16001792.hg.1 | ENST00000008180.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001792.hg.1 | ENST00000396890.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001792.hg.1 | ENST00000444393.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001792.hg.1 | ENST00000525643.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001792.hg.1 | ENST00000526464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001792.hg.1 | ENST00000528163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001792.hg.1 | ENST00000548652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001792.hg.1 | NM_001012631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001792.hg.1 | NM_001012634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001792.hg.1 | TC16000099.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001794.hg.1 | ENST00000008180.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001794.hg.1 | ENST00000396890.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001794.hg.1 | ENST00000444393.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001794.hg.1 | ENST00000525643.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001794.hg.1 | ENST00000526464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001794.hg.1 | ENST00000528163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001794.hg.1 | ENST00000529550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001794.hg.1 | ENST00000548652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001794.hg.1 | NM_001012631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001794.hg.1 | NM_001012634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001794.hg.1 | TC16000099.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001788.hg.1 | ENST00000444393.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001788.hg.1 | ENST00000525643.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001788.hg.1 | ENST00000526464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001788.hg.1 | ENST00000528163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001788.hg.1 | NM_001012631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001788.hg.1 | TC16000099.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001789.hg.1 | ENST00000008180.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001789.hg.1 | ENST00000444393.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001789.hg.1 | ENST00000525643.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001789.hg.1 | ENST00000526464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001789.hg.1 | ENST00000528163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001789.hg.1 | NM_001012631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001789.hg.1 | TC16000099.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001790.hg.1 | ENST00000008180.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001790.hg.1 | ENST00000444393.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001790.hg.1 | ENST00000525643.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001790.hg.1 | ENST00000526464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001790.hg.1 | ENST00000528163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001790.hg.1 | NM_001012631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001790.hg.1 | NM_001012634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001790.hg.1 | TC16000099.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | ENST00000008180.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | ENST00000396890.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | ENST00000444393.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | ENST00000525643.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | ENST00000526464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | ENST00000528163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | ENST00000529550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | ENST00000530538.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | ENST00000548652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | ENST00000549213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | NM_001012631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | NM_001012634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001797.hg.1 | TC16000099.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001791.hg.1 | ENST00000008180.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001791.hg.1 | ENST00000396890.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001791.hg.1 | ENST00000444393.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001791.hg.1 | ENST00000525643.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001791.hg.1 | ENST00000526464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001791.hg.1 | ENST00000528163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001791.hg.1 | NM_001012631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001791.hg.1 | NM_001012634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001791.hg.1 | TC16000099.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001793.hg.1 | ENST00000008180.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001793.hg.1 | ENST00000396890.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001793.hg.1 | ENST00000444393.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001793.hg.1 | ENST00000525643.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001793.hg.1 | ENST00000526464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001793.hg.1 | ENST00000528163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001793.hg.1 | ENST00000548652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001793.hg.1 | NM_001012631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001793.hg.1 | NM_001012634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001793.hg.1 | TC16000099.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | ENST00000008180.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | ENST00000396890.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | ENST00000444393.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | ENST00000525643.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | ENST00000526464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | ENST00000528163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | ENST00000529550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | ENST00000530538.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | ENST00000548652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | NM_001012631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | NM_001012634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001795.hg.1 | TC16000099.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | ENST00000008180.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | ENST00000396890.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | ENST00000444393.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | ENST00000525643.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | ENST00000526464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | ENST00000528163.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | ENST00000529550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | ENST00000530538.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | ENST00000548652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | ENST00000549213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | NM_001012631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | NM_001012634.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001796.hg.1 | TC16000099.hg.1 | HTA TC |

IL32
Genomic Map