Assay Details
| Gene Symbol: RPL37A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000998.4 | NP_000989.1 | 2 - 3 | 209 | 97 | |
| GenBank mRNA | AK289472.1 | - | 2 - 3 | 162 | 97 | |
| AK291857.1 | - | 2 - 3 | 163 | 97 | ||
| BC000555.2 | - | 1 - 2 | 128 | 97 | ||
| BC014262.2 | - | 2 - 3 | 146 | 4654029 | 97 | |
| BC016748.2 | - | 2 - 3 | 162 | 4831278 | 97 | |
| BC063476.1 | - | 2 - 3 | 151 | 5222439 | 97 | |
| BC067789.1 | - | 2 - 3 | 162 | 5299374 | 97 | |
| CR542152.1 | - | 1 - 2 | 131 | 97 | ||
| L06499.1 | - | 2 - 3 | 148 | 97 | ||
| L22154.1 | - | 2 - 3 | 162 | 97 | ||
| X66699.1 | - | 1 - 2 | 135 | 97 | ||
More Information
Set Membership: |
|

RPL37A
Genomic Map