| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800156584.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800156584.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800156584.hg.1 |
TC1800008295.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800156584.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800156584.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
ENST00000579713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008295.hg.1 |
NR_110796.1 |
Refseq |