Assay Details
| Gene Symbol: LOC101928722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011519280.2 | XP_011517582.1 | 8 - 9 | 913 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized locus ESP33 | ESP33 | -3.6kb | Anti-sense | Chr. 9 - 136249883 - 136251845 on NCBI Build GRCh38 | |
| Downstream | coiled-coil domain containing 187 | CCDC187 | 9.7kb | Sense | Chr. 9 - 136267437 - 136306901 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928722
Genomic Map