Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | molybdenum cofactor synthesis 3 | MOCS3 | UBA4 | -38.6kb | Sense | Chr. 20 - 50958814 - 50961863 on NCBI Build GRCh38 |
| Downstream | nuclear factor of activated T-cells 2 | NFATC2 |
NFAT1, NFATP |
377.1kb | Anti-sense | Chr. 20 - 51386957 - 51562857 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map