Assay Details
| Gene Symbol: WDR26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB586698.1 | - | 1 | 219 | 87 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | F03205.1 | - | Chr. 1 - 34596282 - 34596298 [+] on Build GRCh38 | 87 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4742 | MIR4742 | mir-4742 | -19.8kb | Sense | Chr. 1 - 224398227 - 224398311 on NCBI Build GRCh38 |
| Downstream | cornichon family AMPA receptor auxiliary protein 3 | CNIH3 | CNIH-3 | 16.4kb | Anti-sense | Chr. 1 - 224434640 - 224740549 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

WDR26
Genomic Map