Assay Details
| Gene Symbol: C2orf44-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_C2orf44-ALK_C4A20.0 | - | 92 | 98 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf44-ALK fusion| Gene Symbol: C2orf44-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_C2orf44-ALK_C4A20.0 | - | 92 | 98 | ||