Assay Details
| Gene Symbol: PRKAR1A-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PRKAR1A-ALK_P2A20_NGS.0 | - | 62 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRKAR1A-ALK fusion| Gene Symbol: PRKAR1A-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PRKAR1A-ALK_P2A20_NGS.0 | - | 62 | 105 | ||
Set Membership: |
|