Assay Details
| Gene Symbol: NCOA1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NCOA1-ALK_N21A1_NGS.0 | - | 61 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NCOA1-ALK fusion| Gene Symbol: NCOA1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NCOA1-ALK_N21A1_NGS.0 | - | 61 | 97 | ||