Assay Details
| Gene Symbol: FGFR2-CIT fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-CIT_F17C23.0 | - | 89 | 87 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR2-CIT fusion| Gene Symbol: FGFR2-CIT fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-CIT_F17C23.0 | - | 89 | 87 | ||