Assay Details
| Gene Symbol: FGFR2-OFD1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-OFD1_F17O3.0 | - | 52 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR2-OFD1 fusion| Gene Symbol: FGFR2-OFD1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-OFD1_F17O3.0 | - | 52 | 97 | ||