Assay Details
| Gene Symbol: ACTG2-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ACTG2-ALK_A2A18.0 | - | 48 | 102 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACTG2-ALK fusion| Gene Symbol: ACTG2-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ACTG2-ALK_A2A18.0 | - | 48 | 102 | ||
Set Membership: |
|