Assay Details
| Gene Symbol: VCL-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_VCL-ALK_V16A20_COSF1057.0 | - | 69 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VCL-ALK fusion| Gene Symbol: VCL-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_VCL-ALK_V16A20_COSF1057.0 | - | 69 | 78 | ||