Assay Details
| Gene Symbol: EML4-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB663645.1 | - | 1622 | 155 | ||
| OM_EML4-ALK_E14A20_COSF1064.0 | - | 47 | 97 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EML4-ALK fusion| Gene Symbol: EML4-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB663645.1 | - | 1622 | 155 | ||
| OM_EML4-ALK_E14A20_COSF1064.0 | - | 47 | 97 | |||
Set Membership: |
|