Assay Details
| Gene Symbol: ERC1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ERC1_ELKS-RET_E11R12_COSF1508.0 | - | 57 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERC1 fusion| Gene Symbol: ERC1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ERC1_ELKS-RET_E11R12_COSF1508.0 | - | 57 | 89 | ||