Assay Details
| Gene Symbol: KIF5B-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KIF5B-RET_K24R8_COSF1236.0 | - | 75 | 79 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIF5B-RET fusion| Gene Symbol: KIF5B-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KIF5B-RET_K24R8_COSF1236.0 | - | 75 | 79 | ||