Assay Details
| Gene Symbol: ERC1-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ERC1-RET_E12R12.0 | - | 61 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERC1-RET fusion| Gene Symbol: ERC1-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ERC1-RET_E12R12.0 | - | 61 | 84 | ||