Assay Details
| Gene Symbol: SDC4-ROS1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB795241.1 | - | 451 | 78 | ||
| OM_SDC4-ROS1_S4R32_COSF1278.0 | - | 47 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SDC4-ROS1 fusion| Gene Symbol: SDC4-ROS1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB795241.1 | - | 451 | 78 | ||
| OM_SDC4-ROS1_S4R32_COSF1278.0 | - | 47 | 78 | |||