Assay Details
| Gene Symbol: TFG-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TFG-NTRK1_T5N10.0 | - | 65 | 91 | ||
| X85960.1 | - | 617 | 91 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TFG-NTRK1 fusion| Gene Symbol: TFG-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TFG-NTRK1_T5N10.0 | - | 65 | 91 | ||
| X85960.1 | - | 617 | 91 | |||