Assay Details
| Gene Symbol: NFASC-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NFASC-NTRK1_N18N10.0 | - | 100 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NFASC-NTRK1 fusion| Gene Symbol: NFASC-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_NFASC-NTRK1_N18N10.0 | - | 100 | 86 | ||