Assay Details
| Gene Symbol: KIF5B-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB795238.1 | - | 2117 | 86 | ||
| OM_KIF5B-ALK_K17A20_COSF1257.0 | - | 87 | 86 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIF5B-ALK fusion| Gene Symbol: KIF5B-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB795238.1 | - | 2117 | 86 | ||
| OM_KIF5B-ALK_K17A20_COSF1257.0 | - | 87 | 86 | |||