Assay Details
| Gene Symbol: TPM3-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TPM3-ALK_T7A20.0 | - | 93 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPM3-ALK fusion| Gene Symbol: TPM3-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TPM3-ALK_T7A20.0 | - | 93 | 92 | ||