Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | STK24 antisense RNA 1 | STK24-AS1 | -99.0kb | Sense | Chr. 13 - 98577585 - 98578842 on NCBI Build GRCh38 | |
| Downstream | dedicator of cytokinesis 9 | DOCK9 |
ZIZ1, ZIZIMIN1 |
65.7kb | Anti-sense | Chr. 13 - 98793487 - 99086406 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map