| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012055.hg.1 |
ENST00000444217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012055.hg.1 |
ENST00000444217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012055.hg.1 |
NR_046730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012055.hg.1 |
NR_046730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187760.hg.1 |
ENST00000444217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187760.hg.1 |
NR_046730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187760.hg.1 |
ENST00000444217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187760.hg.1 |
NR_046730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187760.hg.1 |
TC0200012055.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200187757.hg.1 |
ENST00000444217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187757.hg.1 |
NR_046730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187757.hg.1 |
ENST00000444217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187757.hg.1 |
NR_046730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187757.hg.1 |
TC0200012055.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200012055.hg.1 |
ENST00000444217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012055.hg.1 |
ENST00000444217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012055.hg.1 |
NR_046730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012055.hg.1 |
NR_046730.1 |
Refseq |