Assay Details
| Gene Symbol: MAJIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300803.1 | NP_001287732.1 | 5 - 6 | 516 | 168 | |
| NM_001318808.1 | NP_001305737.1 | 8 - 9 | 1464 | 168 | ||
| XM_005273918.2 | XP_005273975.1 | 8 - 9 | 744 | 168 | ||
| XM_006718502.2 | XP_006718565.1 | 8 - 9 | 861 | 168 | ||
| XM_006718503.3 | XP_006718566.1 | 8 - 9 | 747 | 168 | ||
| XM_006718505.2 | XP_006718568.1 | 5 - 6 | 627 | 168 | ||
| XM_011544940.2 | XP_011543242.1 | 8 - 9 | 742 | 168 | ||
| XM_011544941.2 | XP_011543243.1 | 2 - 3 | 443 | 168 | ||
| GenBank mRNA | AK093130.1 | - | 8 - 9 | 1458 | 168 | |
| BC033501.1 | - | 5 - 6 | 448 | 168 | ||
| LC068587.1 | - | 6 - 7 | 588 | 168 | ||
More Information
Set Membership: |
|

MAJIN
Genomic Map