Assay Details
| Gene Symbol: MAJIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037225.2 | NP_001032302.1 | 10 - 11 | 928 | 110 | |
| NM_001300803.1 | NP_001287732.1 | 8 - 9 | 758 | 110 | ||
| NM_001318808.1 | NP_001305737.1 | 11 - 12 | 1706 | 110 | ||
| XM_005273918.2 | XP_005273975.1 | 11 - 12 | 986 | 110 | ||
| XM_006718502.2 | XP_006718565.1 | 11 - 12 | 1103 | 110 | ||
| XM_006718503.3 | XP_006718566.1 | 11 - 12 | 989 | 110 | ||
| XM_006718504.2 | XP_006718567.1 | 10 - 11 | 1039 | 110 | ||
| XM_006718505.2 | XP_006718568.1 | 8 - 9 | 869 | 110 | ||
| XM_011544940.2 | XP_011543242.1 | 11 - 12 | 984 | 110 | ||
| XM_011544941.2 | XP_011543243.1 | 5 - 6 | 685 | 110 | ||
| GenBank mRNA | AK093130.1 | - | 11 - 12 | 1700 | 110 | |
| BC033501.1 | - | 8 - 9 | 690 | 110 | ||
| BC106951.2 | - | 9 - 10 | 723 | 40035116 | 110 | |
| BC106952.1 | - | 9 - 10 | 723 | 40035117 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000532617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000532617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | ENST00000532617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185693.hg.1 | TC1100011196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100185691.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185691.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185691.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185691.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185691.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185691.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185691.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185691.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185691.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185691.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185691.hg.1 | TC1100011196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000532617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000532617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | ENST00000532617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011196.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | ENST00000301896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | ENST00000432175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | ENST00000530287.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | ENST00000530444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | ENST00000530735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | NM_001037225.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | NM_001300803.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100185694.hg.1 | TC1100011196.hg.1 | Clariom D TC |

MAJIN
Genomic Map