Assay Details
Gene Symbol: C11orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300913.1 | NP_001287842.1 | 3 - 4 | 521 | 89 | |
NM_022338.3 | NP_071733.1 | 3 - 4 | 521 | 89 | ||
XM_005274053.3 | XP_005274110.1 | 3 - 4 | 696 | 89 | ||
GenBank mRNA | AF264781.1 | - | 3 - 4 | 481 | 89 | |
AF370372.1 | - | 2 - 3 | 89 | |||
AY358754.1 | - | 1 - 2 | 83 | 89 | ||
BC011765.2 | - | 2 - 3 | 370 | 3614861 | 89 | |
BG701401.1 | - | 3 - 4 | 526 | 89 | ||
BI862360.1 | - | 3 - 4 | 495 | 89 | ||
BX384793.2 | - | 2 - 3 | 323 | 89 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002005.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11024076.hg.1 | ENST00000304271.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024076.hg.1 | ENST00000530166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024076.hg.1 | ENST00000533310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024076.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11024076.hg.1 | TC11002005.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11024072.hg.1 | ENST00000304271.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024072.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11024072.hg.1 | TC11002005.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11024073.hg.1 | ENST00000304271.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024073.hg.1 | ENST00000533310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024073.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11024073.hg.1 | TC11002005.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000529339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000529339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000529339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000529339.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000529590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000529590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000529590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000529590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000529590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000532534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000532534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000532534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000532534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000532969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000532969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000532969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11024074.hg.1 | ENST00000304271.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024074.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11024074.hg.1 | TC11002005.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11024075.hg.1 | ENST00000304271.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024075.hg.1 | ENST00000530166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024075.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11024075.hg.1 | TC11002005.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100187788.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187788.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187788.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187788.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187788.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187788.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187788.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187788.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187788.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187788.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187788.hg.1 | TC1100011375.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000529339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000529339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000529339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000529339.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000529590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000529590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000529590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000529590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000529590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000532534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000532534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000532534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000532534.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000532969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000532969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000532969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011375.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187785.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187785.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187785.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187785.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187785.hg.1 | ENST00000531745.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187785.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187785.hg.1 | TC1100011375.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100187792.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187792.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187792.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187792.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187792.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187792.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187792.hg.1 | ENST00000527280.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187792.hg.1 | ENST00000533310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187792.hg.1 | NM_001300913.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187792.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187792.hg.1 | TC1100011375.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100187786.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187786.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187786.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187786.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187786.hg.1 | TC1100011375.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100187787.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187787.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187787.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187787.hg.1 | ENST00000304271.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187787.hg.1 | ENST00000530166.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187787.hg.1 | NM_022338.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187787.hg.1 | TC1100011375.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 162862 | - | - |
