Assay Details
| Gene Symbol: CIB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300922.1 | NP_001287851.1 | 1 - 2 | 97 | 86 | |
| GenBank mRNA | BC069428.1 | - | 1 - 2 | 47 | 7262131 | 86 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181238.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181238.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181238.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181238.hg.1 | TC1900009906.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900181238.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181238.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181238.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | TC1900009906.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181237.hg.1 | NM_054113.3 | Refseq |

CIB3
Genomic Map