Assay Details
| Gene Symbol: CIB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300922.1 | NP_001287851.1 | 3 - 4 | 446 | 142 | |
| NM_054113.3 | NP_473454.1 | 5 - 6 | 593 | 142 | ||
| GenBank mRNA | AB050868.1 | - | 5 - 6 | 543 | 142 | |
| AW295492.1 | - | 1 - 2 | 142 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001264.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19017761.hg.1 | ENST00000269878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017761.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017761.hg.1 | TC19001264.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017764.hg.1 | ENST00000269878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017764.hg.1 | ENST00000379859.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017764.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017764.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017764.hg.1 | TC19001264.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | TC1900009906.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181230.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000379859.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000541493.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | ENST00000597251.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009906.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181227.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181227.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181227.hg.1 | NM_054113.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181227.hg.1 | TC1900009906.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900181227.hg.1 | ENST00000269878.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900181227.hg.1 | NM_001300922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900181227.hg.1 | NM_054113.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 122872 | - | - |

CIB3
Genomic Map